3-Fluor-DL-phenylalanin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-(3-fluorophenyl)propionic acid, H-DL-Phe(3-F)-OH

PC4148-250MG PC4148-5G PC4148-25G
APOSPC4148-250MGEA 21.9 EUR
APOSPC4148-250MG APOSPC4148-5G APOSPC4148-25G
3-Fluor-DL-phenylalanin 98%
3-Fluor-DL-phenylalanin
Formel: C₉H₁₀FNO₂
Molecular Weight: 183,2 g/mol
Schmelzpunkt: 240…250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004273
CAS-Nummer: 456-88-2
EINECS: 220-104-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR