Sodium 3-fluoro-2-oxopropanoate monohydrate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-2-oxopropanoic acid sodium salt monohydrate, 3-Fluoropyruvic acid sodium salt monohydrate

PC4205-1G PC4205-5G
APOSPC4205-1GEA 434 EUR
APOSPC4205-1G APOSPC4205-5G
Sodium 3-fluoro-2-oxopropanoate monohydrate 98%
Sodium 3-fluoro-2-oxopropanoate monohydrate
Formel: C₃H₂FO₃·Na·H₂O
Molecular Weight: 146,05 g/mol
Schmelzpunkt: 145 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00149108
CAS-Nummer: 238754-68-2
EINECS: 220-880-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR