2-Bromo-6-fluoro-nicotinic acid 95%
Lieferant: Apollo Scientific
Synonyme:
2-Bromo-6-fluoronicotinic acid
PC421066-1G
PC421066-250MG
PC421066-5G
APOSPC421066-1GEA
491
EUR
APOSPC421066-1G
APOSPC421066-250MG
APOSPC421066-5G
2-Bromo-6-fluoro-nicotinic acid 95%
2-Bromo-6-fluoro-nicotinic acid
Formel:
C₆H₃BrFNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185798 CAS-Nummer: 1214332-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...