2-Amino-4-methoxy-5-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

Synonyme: 4-Methoxy-5-(trifluoromethyl)pyridin-2-amine

PC421075-100MG PC421075-1G PC421075-250MG
APOSPC421075-100MGEA 74.1 EUR
APOSPC421075-100MG APOSPC421075-1G APOSPC421075-250MG
2-Amino-4-methoxy-5-(trifluoromethyl)pyridine
2-Amino-4-methoxy-5-(trifluoromethyl)pyridine
Formel: C₇H₇F₃N₂O
Molecular Weight: 192,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16606208
CAS-Nummer: 1227571-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR