2,6-Dichloro-3-fluoroisonicotinic acid 95%

Lieferant: Apollo Scientific
Achtung

PC421103-1G PC421103-250MG
APOSPC421103-1GEA 475 EUR
APOSPC421103-1G APOSPC421103-250MG
2,6-Dichloro-3-fluoroisonicotinic acid 95%
2,6-Dichloro-3-fluoroisonicotinic acid
Formel: C₆H₂Cl₂FNO₂
Molecular Weight: 209,99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24490307
CAS-Nummer: 149468-00-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR