5-Bromo-1-(difluoromethyl)pyridin-2(1h)-one 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-1-(difluoromethyl)pyridin-2(1H)-one

PC421161-1G PC421161-250MG PC421161-5G
APOSPC421161-1GEA 207 EUR
APOSPC421161-1G APOSPC421161-250MG APOSPC421161-5G
5-Bromo-1-(difluoromethyl)pyridin-2(1h)-one 95+%
5-Bromo-1-(difluoromethyl)pyridin-2(1h)-one
Formel: C₆H₄BrF₂NO
Molecular Weight: 224,003 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12911848
CAS-Nummer: 832735-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR