2,6-Dichloro-4-fluoroiodobenzene 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Dichloro-5-fluoro-2-iodobenzene

PC421169-250MG PC421169-1G
APOSPC421169-250MGEA 94.4 EUR
APOSPC421169-250MG APOSPC421169-1G
2,6-Dichloro-4-fluoroiodobenzene 95%
2,6-Dichloro-4-fluoroiodobenzene
Formel: C₆H₂Cl₂FI
Molecular Weight: 290,889 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194304
CAS-Nummer: 939990-10-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR