(1R,2R)-2-(3,4-Difluorophenyl)cyclopropanecarboxylic acid 97%

Lieferant: Apollo Scientific

PC421187-100MG PC421187-250MG
APOSPC421187-100MGEA 701 EUR
APOSPC421187-100MG APOSPC421187-250MG
(1R,2R)-2-(3,4-Difluorophenyl)cyclopropanecarboxylic acid 97%
(1R,2R)-2-(3,4-Difluorophenyl)cyclopropanecarboxylic acid
Formel: C₁₀H₈F₂O₂
Molecular Weight: 198.166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20639702
CAS-Nummer: 220352-36-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR