(2-Amino-3-fluorophenyl)(phenyl)methanone 95%

Lieferant: Apollo Scientific

PC421188-100MG PC421188-250MG
APOSPC421188-100MGEA 593 EUR
APOSPC421188-100MG APOSPC421188-250MG
(2-Amino-3-fluorophenyl)(phenyl)methanone 95%
(2-Amino-3-fluorophenyl)(phenyl)methanone
Formel: C₁₃H₁₀FNO
Molecular Weight: 215,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27836815
CAS-Nummer: 1584139-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR