(4-(5-Fluoropyrimidin-2-yl)phenyl)methanol 95%

Lieferant: Apollo Scientific

PC421189-100MG PC421189-1G PC421189-250MG
APOSPC421189-100MGEA 360 EUR
APOSPC421189-100MG APOSPC421189-1G APOSPC421189-250MG
(4-(5-Fluoropyrimidin-2-yl)phenyl)methanol 95%
(4-(5-Fluoropyrimidin-2-yl)phenyl)methanol
Formel: C₁₁H₉FN₂O
Molecular Weight: 204.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20441490
CAS-Nummer: 1139432-29-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR