(Z)-3-Chloro-3-(3-fluorophenyl)acrylonitrile 98%
Lieferant: Apollo Scientific
PC421195-100MG
PC421195-1G
PC421195-250MG
APOSPC421195-100MGEA
316
EUR
APOSPC421195-100MG
APOSPC421195-1G
APOSPC421195-250MG
(Z)-3-Chloro-3-(3-fluorophenyl)acrylonitrile 98%
(Z)-3-Chloro-3-(3-fluorophenyl)acrylonitrile
Formel:
C₉H₅ClFN Molecular Weight: 181,594 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11207883 CAS-Nummer: 1192875-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...