5-Fluoro-1-methyl-6-oxo-1,6-dihydropyridine-2-carbonitrile

Lieferant: Apollo Scientific
Achtung

PC421223-1G PC421223-250MG
APOSPC421223-1GEA 1010 EUR
APOSPC421223-1G APOSPC421223-250MG
5-Fluoro-1-methyl-6-oxo-1,6-dihydropyridine-2-carbonitrile
5-Fluoro-1-methyl-6-oxo-1,6-dihydropyridine-2-carbonitrile
Formel: C₇H₅FN₂O
Molecular Weight: 152.126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29126235
CAS-Nummer: 1887015-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR