6-Fluoro-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one 98%
Lieferant: Apollo Scientific
PC421226-100MG
PC421226-250MG
APOSPC421226-100MGEA
165
EUR
APOSPC421226-100MG
APOSPC421226-250MG
6-Fluoro-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one 98%
6-Fluoro-4,5-dihydro-1H-benzo[b]azepin-2(3H)-one
Formel:
C₁₀H₁₀FNO Molecular Weight: 179,191 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11977761 CAS-Nummer: 145485-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...