S-Methyl 2,5-difluorobenzothioate 98%
Lieferant: Apollo Scientific
PC421233-100MG PC421233-250MG
APOSPC421233-100MGEA 691 EUR
APOSPC421233-100MG APOSPC421233-250MG
S-Methyl 2,5-difluorobenzothioate 98%
S-Methyl 2,5-difluorobenzothioate
Formel: C₈H₆F₂OS
Molecular Weight: 188.194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23160076
CAS-Nummer: 1146210-64-1