3-Fluoro-5-nitrobenzyl chloride 95%

Lieferant: Apollo Scientific

Synonyme: 1-(Chloromethyl)-3-fluoro-5-nitrobenzene

PC430000-5G
APOSPC430000-5GEA 87 EUR
APOSPC430000-5G
3-Fluoro-5-nitrobenzyl chloride 95%
3-Fluoro-5-nitrobenzyl chloride
Formel: C₇H₅ClFNO₂
Molecular Weight: 189,572 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185626
CAS-Nummer: 1214344-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR