2-(Trifluoromethoxy)ethylamine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Trifluoromethoxy)ethanamine hydrochloride

PC430117-1G PC430117-5G
APOSPC430117-1GEA 260 EUR
APOSPC430117-1G APOSPC430117-5G
2-(Trifluoromethoxy)ethylamine hydrochloride 97%
2-(Trifluoromethoxy)ethylamine hydrochloride
Formel: C₃H₆F₃NO·ClH
Molecular Weight: 165,542 g/mol
Schmelzpunkt: 189…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08447280
CAS-Nummer: 886050-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR