Potassium cyclohexylmethyl trifluoroborate
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Potassium (cyclohexylmethyl)trifluoroborate
PC430518-500MG
PC430518-1G
PC430518-5G
APOSPC430518-500MGEA
0
EUR
APOSPC430518-500MG
APOSPC430518-1G
APOSPC430518-5G
Potassium cyclohexylmethyl trifluoroborate
Potassium cyclohexylmethyl trifluoroborate
Formel:
C₇H₁₃BF₃·K Molecular Weight: 204,083 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11052658 CAS-Nummer: 1346647-18-4 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...