3-Fluoro-4-nitrobenzotrifluoride 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-4-(trifluoromethyl)nitrobenzene, 2-Fluoro-1-nitro-4-(trifluoromethyl)benzene
PC4393-1G
PC4393-5G
PC4393-10G
APOSPC4393-1GEA
30.6
EUR
APOSPC4393-1G
APOSPC4393-5G
APOSPC4393-10G
3-Fluoro-4-nitrobenzotrifluoride 97%
3-Fluoro-4-nitrobenzotrifluoride
Formel:
C₇H₃F₄NO₂ Molecular Weight: 209.098 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840208 CAS-Nummer: 402-12-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...