5-Fluoro-2,3-diaminopyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoropyridine-2,3-diamine

PC445029-1G PC445029-5G
APOSPC445029-1GEA 115 EUR
APOSPC445029-1G APOSPC445029-5G
5-Fluoro-2,3-diaminopyridine
5-Fluoro-2,3-diaminopyridine
Formel: C₅H₆FN₃
Molecular Weight: 127,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08062986
CAS-Nummer: 212268-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR