4-Bromo-2-chloro-5-fluoropyridine 97%

Lieferant: Apollo Scientific

PC445056-250MG PC445056-1G PC445056-5G
APOSPC445056-250MGEA 77.1 EUR
APOSPC445056-250MG APOSPC445056-1G APOSPC445056-5G
4-Bromo-2-chloro-5-fluoropyridine 97%
4-Bromo-2-chloro-5-fluoropyridine
Formel: C₅H₂BrClFN
Molecular Weight: 210.431 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659500
CAS-Nummer: 884495-10-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR