N-[3-Bromo-6-(morpholin-4-yl)-4-(trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine

Lieferant: Apollo Scientific

PC446020-1G PC446020-5G
APOSPC446020-1GEA 308 EUR
APOSPC446020-1G APOSPC446020-5G
N-[3-Bromo-6-(morpholin-4-yl)-4-(trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine
N-[3-Bromo-6-(morpholin-4-yl)-4-(trifluoromethyl)pyridin-2-yl]ethane-1,2-diamine
Formel: C₁₂H₁₆BrF₃N₄O
Molecular Weight: 369,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052388
CAS-Nummer: 1089330-42-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR