2-Fluoro-pyridine-3-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

PC446034-1G PC446034-250MG
APOSPC446034-1GEA 383 EUR
APOSPC446034-1G APOSPC446034-250MG
2-Fluoro-pyridine-3-sulphonyl chloride
2-Fluoro-pyridine-3-sulphonyl chloride
Formel: C₅H₃ClFNO₂S
Molecular Weight: 195.599 g/mol
Siedepunkt: 96…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11227180
CAS-Nummer: 1089330-70-0
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR