(3,5-Difluoropyridin-2-yl)methanol 95+%

Lieferant: Apollo Scientific

PC446167-1G PC446167-5G
APOSPC446167-1GEA 122 EUR
APOSPC446167-1G APOSPC446167-5G
(3,5-Difluoropyridin-2-yl)methanol 95+%
(3,5-Difluoropyridin-2-yl)methanol
Formel: C₆H₅F₂NO
Molecular Weight: 145,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11520700
CAS-Nummer: 1065267-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR