2-Bromo-3-(difluoromethoxy)pyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC448004-1G
PC448004-250MG
PC448004-5G
APOSPC448004-1GEA
191
EUR
APOSPC448004-1G
APOSPC448004-250MG
APOSPC448004-5G
2-Bromo-3-(difluoromethoxy)pyridine 95%
2-Bromo-3-(difluoromethoxy)pyridine
Formel:
C₆H₄BrF₂NO Molecular Weight: 224,003 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185840 CAS-Nummer: 947249-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...