4-Bromo-1-cyclopentyl-3-(trifluoromethyl)pyrazole

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-1-cyclopentyl-3-(trifluoromethyl)-1H-pyrazole

PC448007-1G PC448007-25G PC448007-5G
APOSPC448007-1GEA 87 EUR
APOSPC448007-1G APOSPC448007-25G APOSPC448007-5G
4-Bromo-1-cyclopentyl-3-(trifluoromethyl)pyrazole
4-Bromo-1-cyclopentyl-3-(trifluoromethyl)pyrazole
Formel: C₉H₁₀BrF₃N₂
Molecular Weight: 283,088 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24448783
CAS-Nummer: 1437794-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR