3-(Difluoromethoxy)pyridin-2-amine 95%

Lieferant: Apollo Scientific
Achtung

PC448010-1G PC448010-250MG
APOSPC448010-1GEA 125 EUR
APOSPC448010-1G APOSPC448010-250MG
3-(Difluoromethoxy)pyridin-2-amine 95%
3-(Difluoromethoxy)pyridin-2-amine
Formel: C₆H₆F₂N₂O
Molecular Weight: 160.121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12405788
CAS-Nummer: 947249-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR