7-Bromo-5-fluoro-1H-Benzotriazole 95%
Lieferant: Apollo Scientific
Synonyme:
4-Bromo-6-fluoro-1H-benzo[d][1,2,3]triazole
PC448209-1G
PC448209-100MG
PC448209-250MG
APOSPC448209-1GEA
519
EUR
APOSPC448209-1G
APOSPC448209-100MG
APOSPC448209-250MG
7-Bromo-5-fluoro-1H-Benzotriazole 95%
7-Bromo-5-fluoro-1H-Benzotriazole
Formel:
C₆H₃BrFN₃ Molecular Weight: 216,011 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29127456 CAS-Nummer: 937013-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...