2-Chloro-3,5-difluorobenzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Chloro-3,5-difluorophenyl)methanol

PC44896-1G PC44896-250MG PC44896-5G
APOSPC44896-1GEA 58 EUR
APOSPC44896-1G APOSPC44896-250MG APOSPC44896-5G
2-Chloro-3,5-difluorobenzyl alcohol 95%
2-Chloro-3,5-difluorobenzyl alcohol
Formel: C₇H₅ClF₂O
Molecular Weight: 178.564 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28520352
CAS-Nummer: 1785519-54-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR