1-(4-Fluorophenyl)cyclopropylmethanol
Lieferant: Apollo Scientific
|
Achtung
|
PC449014-1G
PC449014-250MG
APOSPC449014-1GEA
478
EUR
APOSPC449014-1G
APOSPC449014-250MG
1-(4-Fluorophenyl)cyclopropylmethanol
1-(4-Fluorophenyl)cyclopropylmethanol
Formel:
C₁₀H₁₁FO Molecular Weight: 166,192 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07374455 CAS-Nummer: 198976-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...