2-Chloro-4,6-difluorobenzaldehyde 95%
Lieferant: Apollo Scientific
PC44902-1G
PC44902-250MG
PC44902-5G
APOSPC44902-1GEA
128
EUR
APOSPC44902-1G
APOSPC44902-250MG
APOSPC44902-5G
2-Chloro-4,6-difluorobenzaldehyde 95%
2-Chloro-4,6-difluorobenzaldehyde
Formel:
C₇H₃ClF₂O Molecular Weight: 176,548 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18415553 CAS-Nummer: 1261493-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...