1-Ethyl-3-(trifluoromethyl)benzene

Lieferant: Apollo Scientific
Achtung

PC450011-1G PC450011-250MG
APOSPC450011-1GEA 799 EUR
APOSPC450011-1G APOSPC450011-250MG
1-Ethyl-3-(trifluoromethyl)benzene
1-Ethyl-3-(trifluoromethyl)benzene
Formel: C₉H₉F₃
Molecular Weight: 174,163 g/mol
Siedepunkt: 52 °C (6 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD24842561
CAS-Nummer: 27190-70-1
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR