(1-Fluoroprop-1-en-1-yl)benzene

Lieferant: Apollo Scientific

PC450040-100MG PC450040-250MG
APOSPC450040-100MGEA 139 EUR
APOSPC450040-100MG APOSPC450040-250MG
(1-Fluoroprop-1-en-1-yl)benzene
(1-Fluoroprop-1-en-1-yl)benzene
Formel: C₉H₉F
Molecular Weight: 136,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22377929
CAS-Nummer: 75321-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR