Hexakis(3-fluoropropoxy)phosphazene
Lieferant: Apollo Scientific
PC450048-1G
PC450048-250MG
APOSPC450048-1GEA
1070
EUR
APOSPC450048-1G
APOSPC450048-250MG
Hexakis(3-fluoropropoxy)phosphazene
Hexakis(3-fluoropropoxy)phosphazene
Formel:
C₁₈H₃₆F₆N₃O₆P₃ Molecular Weight: 597,407 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20489426 CAS-Nummer: 1346521-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...