Tris(2,2,3,3,3-pentafluoropropyl)orthoformate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-[Bis(2,2,3,3,3-pentafluoropropoxy)methoxy]-1,1,1,2,2-pentafluoropropane
PC450081-1G
PC450081-5G
APOSPC450081-1GEA
284
EUR
APOSPC450081-1G
APOSPC450081-5G
Tris(2,2,3,3,3-pentafluoropropyl)orthoformate
Tris(2,2,3,3,3-pentafluoropropyl)orthoformate
Formel:
C₁₀H₇F₁₅O₃ Molecular Weight: 460,137 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22123976 CAS-Nummer: 917379-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...