(Trifluoromethanesulphonyl)methane
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Trifluoro(methanesulphonyl)methane
PC450086-1G
PC450086-5G
APOSPC450086-1GEA
285
EUR
APOSPC450086-1G
APOSPC450086-5G
(Trifluoromethanesulphonyl)methane
(Trifluoromethanesulphonyl)methane
Formel:
C₂H₃F₃O₂S Molecular Weight: 148,104 g/mol Siedepunkt: 130 °C Dichte: 1,5141 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD18916802 CAS-Nummer: 421-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...