Perfluorohept-3-ene
Lieferant: Apollo Scientific
|
Achtung
|
PC450098-1G
PC450098-250MG
APOSPC450098-1GEA
458
EUR
APOSPC450098-1G
APOSPC450098-250MG
Perfluorohept-3-ene
Perfluorohept-3-ene
Formel:
C₇F₁₄ Lagertemperatur: Kühlschrank |
MDL:
MFCD28054275 CAS-Nummer: 71039-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...