4-Cyano-2-nitro-5-(trifluoromethyl)trifluoroacetanilide

Lieferant: Apollo Scientific

PC450113-1G PC450113-250MG
APOSPC450113-1GEA 546 EUR
APOSPC450113-1G APOSPC450113-250MG
4-Cyano-2-nitro-5-(trifluoromethyl)trifluoroacetanilide
4-Cyano-2-nitro-5-(trifluoromethyl)trifluoroacetanilide
Formel: C₁₀H₃F₆N₃O₃
Molecular Weight: 327,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18074384
CAS-Nummer: 1155800-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR