Ethyl (2,2,2-trifluoroethyl) carbonate

Lieferant: Apollo Scientific
Achtung

PC450128-1G PC450128-5G
APOSPC450128-1GEA 0 EUR
APOSPC450128-1G APOSPC450128-5G
Ethyl (2,2,2-trifluoroethyl) carbonate
Ethyl (2,2,2-trifluoroethyl) carbonate
Formel: C₅H₇F₃O₃
Molecular Weight: 172,102 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054294
CAS-Nummer: 156783-96-9
UN: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR