4-[(2,2,2-Trifluoroethyl)amino]phenol

Lieferant: Apollo Scientific

PC450131-1G PC450131-5G
APOSPC450131-1GEA 139 EUR
APOSPC450131-1G APOSPC450131-5G
4-[(2,2,2-Trifluoroethyl)amino]phenol
4-[(2,2,2-Trifluoroethyl)amino]phenol
Formel: C₈H₈F₃NO
Molecular Weight: 191,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18074381
CAS-Nummer: 1011805-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR