1-Bromo-1,1-difluoro-2,4-pentanedione

Lieferant: Apollo Scientific

PC450135-1G PC450135-5G
APOSPC450135-1GEA 109 EUR
APOSPC450135-1G APOSPC450135-5G
1-Bromo-1,1-difluoro-2,4-pentanedione
1-Bromo-1,1-difluoro-2,4-pentanedione
Formel: C₅H₅BrF₂O₂
Molecular Weight: 214,993 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18074350
CAS-Nummer: 164342-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR