3,3,3-Trifluoropropyl difluoromethylether

Lieferant: Apollo Scientific

PC450139-1G PC450139-250MG
APOSPC450139-1GEA 227 EUR
APOSPC450139-1G APOSPC450139-250MG
3,3,3-Trifluoropropyl difluoromethylether
3,3,3-Trifluoropropyl difluoromethylether
Formel: C₄H₅F₅O
Molecular Weight: 164,074 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27918472
CAS-Nummer: 913607-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR