1,3,3,4,4,5,5-Heptafluoro-2-methoxycyclopent-1-ene

Lieferant: Apollo Scientific
Achtung

PC450141-1G PC450141-5G
APOSPC450141-1GEA 219 EUR
APOSPC450141-1G APOSPC450141-5G
1,3,3,4,4,5,5-Heptafluoro-2-methoxycyclopent-1-ene
1,3,3,4,4,5,5-Heptafluoro-2-methoxycyclopent-1-ene
Formel: C₆H₃F₇O
Molecular Weight: 224,076 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01735620
CAS-Nummer: 1759-60-0
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR