6-(Trifluoromethoxy)-2-(trifluoromethyl)-1H-benzimidazole

Lieferant: Apollo Scientific

PC450148-1G PC450148-5G
APOSPC450148-1GEA 328 EUR
APOSPC450148-1G APOSPC450148-5G
6-(Trifluoromethoxy)-2-(trifluoromethyl)-1H-benzimidazole
6-(Trifluoromethoxy)-2-(trifluoromethyl)-1H-benzimidazole
Formel: C₉H₄F₆N₂O
Molecular Weight: 270,131 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18074397
CAS-Nummer: 113638-38-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR