Tris(2,2,3,3-tetrafluoropropyl)orthoformate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-[Bis(2,2,3,3-tetrafluoropropoxy)methoxy]-1,1,2,2-tetrafluoropropane
PC450152-5G
PC450152-1G
APOSPC450152-5GEA
460
EUR
APOSPC450152-5G
APOSPC450152-1G
Tris(2,2,3,3-tetrafluoropropyl)orthoformate 97%
Tris(2,2,3,3-tetrafluoropropyl)orthoformate
Formel:
C₁₀H₁₀F₁₂O₃ Molecular Weight: 406,165 g/mol Siedepunkt: 223 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD22123981 CAS-Nummer: 21983-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...