2-(2-Fluorophenyl)-1H-benzimidazole

Lieferant: Apollo Scientific

PC450170-1G PC450170-5G
APOSPC450170-1GEA 183 EUR
APOSPC450170-1G APOSPC450170-5G
2-(2-Fluorophenyl)-1H-benzimidazole
2-(2-Fluorophenyl)-1H-benzimidazole
Formel: C₁₃H₉FN₂
Molecular Weight: 212,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01086283
CAS-Nummer: 321-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR