Methyl 4,4,5,5,6,6,6-heptafluorohexanoate
Lieferant: Apollo Scientific
PC450172-1G
PC450172-5G
APOSPC450172-1GEA
139
EUR
APOSPC450172-1G
APOSPC450172-5G
Methyl 4,4,5,5,6,6,6-heptafluorohexanoate
Methyl 4,4,5,5,6,6,6-heptafluorohexanoate
Formel:
C₇H₇F₇O₂ Molecular Weight: 256,118 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20489375 CAS-Nummer: 2070-60-2 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...