trans-1,2-Dichlorohexafluorocyclobutane 95%

Lieferant: Apollo Scientific
Achtung

PC450187-1G PC450187-5G
APOSPC450187-1GEA 260 EUR
APOSPC450187-1G APOSPC450187-5G
trans-1,2-Dichlorohexafluorocyclobutane 95%
trans-1,2-Dichlorohexafluorocyclobutane
Formel: C₄Cl₂F₆
Molecular Weight: 232.939 g/mol
Dichte: 1.64 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD23704312
CAS-Nummer: 3832-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR