Tris(3-fluoropropyl)borate

Lieferant: Apollo Scientific
Achtung

PC450192-1G PC450192-250MG PC450192-5G
APOSPC450192-1GEA 427 EUR
APOSPC450192-1G APOSPC450192-250MG APOSPC450192-5G
Tris(3-fluoropropyl)borate
Tris(3-fluoropropyl)borate
Formel: C₉H₁₈BF₃O₃
Molecular Weight: 242,044 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20489408
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR