4,4,4-Trifluorobutyl difluoromethylether

Lieferant: Apollo Scientific
Achtung

PC450212-1G PC450212-5G
APOSPC450212-1GEA 145 EUR
APOSPC450212-1G APOSPC450212-5G
4,4,4-Trifluorobutyl difluoromethylether
4,4,4-Trifluorobutyl difluoromethylether
Formel: C₅H₇F₅O
Molecular Weight: 178,1 g/mol
Siedepunkt: 98 °C
Lagertemperatur: Kühlschrank
MDL: MFCD27918476
CAS-Nummer: 1823866-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR