2,2,3,3,4,4,5,5,5-Nonafluoropentyl difluoromethylether 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC450217-1G
PC450217-5G
APOSPC450217-1GEA
857
EUR
APOSPC450217-1G
APOSPC450217-5G
2,2,3,3,4,4,5,5,5-Nonafluoropentyl difluoromethylether 97%
2,2,3,3,4,4,5,5,5-Nonafluoropentyl difluoromethylether
Formel:
C₆H₃F₁₁O Molecular Weight: 300,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27918475 CAS-Nummer: 1661005-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...